Ethyl-2-piperidinecarboxylate 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl 2-Piperidinecarboxylate, Ethyl pipecolinate

L10532-03 L10532-06 L10532-14
CAAAAL10532-03EA 44.71 CAD
CAAAAL10532-03 CAAAAL10532-06 CAAAAL10532-14
Ethyl-2-piperidinecarboxylate 98+%
Ethyl 2-piperidinecarboxylate

Purity: 98+% CAS No.: 15862-72-3 Molecular Formula: C8H15NO2 Formula Weight: 157.21 Flash Point: 46°(114°F) Refractive Index: 1.4560 Boiling Point: 216-217° Beilstein Reference: 81694 UN No.: 1993 MDL No.: MFCD00005980

Formula: C₈H₁₅NO₂
MW: 157.21 g/mol
Boiling Pt: 216…217 °C
Density: 1.006
Flash Pt: 46 °C(114 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005980
CAS Number: 15862-72-3
EINECS: 239-990-6
UN: 1993
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR