Ethyl pentafluorobenzoate 98%

Supplier: Thermo Scientific Chemicals
Warning

L10523-06 L10523-14
CAAAAL10523-06EA 47.4 CAD
CAAAAL10523-06 CAAAAL10523-14
Ethyl pentafluorobenzoate 98%
Ethyl pentafluorobenzoate

Purity: 98% CAS No.: 4522-93-4 Molecular Formula: C9H5F5O2 Formula Weight: 240.13 Flash Point: 80°(176°F) Refractive Index: 1.4280 Boiling Point: 112-114° Beilstein Reference: 2054770 MDL No.: MFCD00039211

Formula: C₉H₅F₅O₂
Boiling Pt: 112…114 °C
Density: 1,431
Flash Pt: 80 °C(176 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039211
CAS Number: 4522-93-4
EINECS: 224-854-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR