Dimethyl adipimidate dihydrochloride 97%

Supplier: Thermo Scientific Chemicals
Warning

L10515-04 L10515-09
CAAAAL10515-04EA 77.1 CAD
CAAAAL10515-04 CAAAAL10515-09
Dimethyl adipimidate dihydrochloride 97%
Dimethyl adipimidate dihydrochloride

Purity: 97% CAS No.: 14620-72-5 Molecular Formula: C8H16N2O2.2HCl Formula Weight: 245.15 Melting Point: ca 215° dec. MDL No.: MFCD00012573

Formula: C₈H₁₆N₂O₂
Melting Pt: ca 215 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012573
CAS Number: 14620-72-5
EINECS: 238-660-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR