Methyl-4-isothiocyanatobenzoate 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-(Methoxycarbonyl)phenyl isothiocyanate

L10474-03 L10474-06
CAAAAL10474-03EA 104.91 CAD
CAAAAL10474-03 CAAAAL10474-06
Methyl-4-isothiocyanatobenzoate 98+%
Methyl 4-isothiocyanatobenzoate

Purity: 98+% CAS No.: 3662-78-0 Molecular Formula: C9H7NO2S Formula Weight: 193.23 Odor: Irritating Melting Point: 46-50° Beilstein Reference: 2100504 UN No.: 2923 MDL No.: MFCD00041099

Formula: C₉H₇NO₂S
MW: 193.23 g/mol
Melting Pt: 46…50 °C
Storage Temperature: Ambient
MDL Number: MFCD00041099
CAS Number: 3662-78-0
EINECS: 240-164-2
UN: 2923
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR