1H,1H-Heptafluorobutyl acrylate 97% stabilized

Supplier: Thermo Scientific Chemicals
Warning


Stabilization: Stabilised with 4-methoxyphenol (MEHQ) 100 ppm

L10400-06 L10400-14
CAAAAL10400-06EA 94.48 CAD
CAAAAL10400-06 CAAAAL10400-14
1H,1H-Heptafluorobutyl acrylate 97% stabilized
1H,1H-Heptafluorobutyl acrylate

Stab. with 100 ppm 4-methoxyphenol


Purity: 97% CAS No.: 424-64-6 Molecular Formula: CH2=CHCO2CH2CF2CF2CF3 Formula Weight: 254.11 Flash Point: 31°(87°F) Refractive Index: 1.3310 Odor: Pungent Boiling Point: 57-58°/30mm Beilstein Reference: 1792520 UN No.: 3272 MDL No.: MFCD00039252

Formula: C₇H₅F₇O₂
MW: 254.11 g/mol
Boiling Pt: 57…58 °C/30 mm
Density: 1.418
Flash Pt: 31 °C(87 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039252
CAS Number: 424-64-6
EINECS: 207-036-8
UN: 3272
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR