2,2,3,3,3-Pentafluoropropyl acrylate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2,2,3,3,3-Pentafluoropropyl prop-2-enoate
L10343-03
L10343-06
CAAAAL10343-03EA
46.14
CAD
CAAAAL10343-03
CAAAAL10343-06
2,2,3,3,3-Pentafluoropropyl acrylate 97%
2,2,3,3,3-Pentafluoropropyl acrylate
Purity: 97%
CAS No.: 356-86-5
Molecular Formula: C6H5F5O2
Formula Weight: 204.10
Storage Temperature: 2°C to 8°C
Flash Point: 23°(73°F)
Refractive Index: 1.3360
Odor: Pungent
Boiling Point: 49-50°/100mm
Beilstein Reference: 1779140
UN No.: 3272
MDL No.: MFCD00039257
Formula:
C₆H₅F₅O₂ Boiling Pt: 49…50 °C/100 mm Density: 1.320 Flash Pt: 23 °C(73 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00039257 CAS Number: 356-86-5 EINECS: 206-608-4 UN: 3272 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...