N-Acetyl-DL-alanine ≥95%
Supplier: Thermo Scientific Chemicals
L10329-06
L10329-14
CAAAAL10329-06EA
42.03
CAD
CAAAAL10329-06
CAAAAL10329-14
N-Acetyl-DL-alanine ≥95%
N-Acetyl-DL-alanine
Purity: 97+%
CAS No.: 1115-69-1
Molecular Formula: C5H9NO3
Formula Weight: 131.13
Storage Temperature: 2°C to 8°C
Melting Point: 135-139°
Beilstein Reference: 774333
MDL No.: MFCD00037238
Formula:
C₅H₉NO₃ Melting Pt: 135…139 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00037238 CAS Number: 1115-69-1 EINECS: 214-229-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...