Tetrahydrofurfuryl acrylate 90% stabilized, tech.
Supplier: Thermo Scientific Chemicals
|
Danger
|
Stabilization:
Stabilised with hydroquinone (HQ) 500 ppm
L10299-18
L10299-30
CAAAAL10299-18EA
59.09
CAD
CAAAAL10299-18
CAAAAL10299-30
Tetrahydrofurfuryl acrylate 90% stabilized, tech.
Tetrahydrofurfuryl acrylate
Stab. with 500ppm hydroquinone
Purity: 90%
CAS No.: 2399-48-6
Molecular Formula: C8H12O3
Formula Weight: 156.18
Flash Point: 93°(199°F)
Refractive Index: 1.4550
Boiling Point: 88-89°/9mm
Beilstein Reference: 113115
UN No.: 3334
MDL No.: MFCD00014516
Formula:
C₈H₁₂O₃ Boiling Pt: 88…89 °C/9 mm Density: 1.030 Flash Pt: 93 °C(199 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014516 CAS Number: 2399-48-6 EINECS: 219-268-7 UN: 3334 ADR: "9",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...