2,5-Thiophenedicarboxylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

L10293-03 L10293-06 L10293-14
CAAAAL10293-03EA 40.13 CAD
CAAAAL10293-03 CAAAAL10293-06 CAAAAL10293-14
2,5-Thiophenedicarboxylic acid 97%
2,5-Thiophenedicarboxylic acid

Purity: 97% CAS No.: 4282-31-9 Molecular Formula: C6H4O4S Formula Weight: 172.16 Melting Point: 355-360° Beilstein Reference: 136781

Formula: C₆H₄O₄S
Melting Pt: 355…360 °C
Storage Temperature: Ambient
MDL Number: MFCD00016896
CAS Number: 4282-31-9
EINECS: 224-284-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR