Ethyl-4,4,4-trifluorocrotonate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ethyl (E)-4,4,4-trifluorobut-2-enoate, Ethyl 4,4,4-trifluorocrotonate
L10285-03
L10285-06
CAAAAL10285-03EA
46.61
CAD
CAAAAL10285-03
CAAAAL10285-06
Ethyl-4,4,4-trifluorocrotonate 98%
Ethyl 4,4,4-trifluorocrotonate
Purity: 98%
CAS No.: 25597-16-4
Molecular Formula: CF3CH=CHCO2CH2CH3
Formula Weight: 168.12
Flash Point: 25°(77°F)
Refractive Index: 1.3601
Boiling Point: 113-115°
Beilstein Reference: 1707262
UN No.: 3272
MDL No.: MFCD00009903
Formula:
C₆H₇F₃O₂ MW: 168.12 g/mol Boiling Pt: 113…115 °C Density: 1.125 Flash Pt: 25 °C (77 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009903 CAS Number: 25597-16-4 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...