4-(Trifluoroacetyl)benzoic acid 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L10242-03
L10242-MD
CAAAAL10242-03EA
170.64
CAD
CAAAAL10242-03
CAAAAL10242-MD
4-(Trifluoroacetyl)benzoic acid 97+%
4-(Trifluoroacetyl)benzoic acid
Purity: 97+%
CAS No.: 58808-59-6
Molecular Formula: C9H5F3O3
Formula Weight: 218.13
Melting Point: 177-179°
Beilstein Reference: 2728778
Formula:
C₉H₅F₃O₃ Melting Pt: 177…179 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052340 CAS Number: 58808-59-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...