10,12-Tricosadiynoic acid 96%

Supplier: Thermo Scientific Chemicals

Synonyms: 10,12-Tricosadiynoicacid

L10231-03 L10231-06
CAAAAL10231-03EA 99.38 CAD
CAAAAL10231-03 CAAAAL10231-06
10,12-Tricosadiynoic acid 96%
10,12-Tricosadiynoic acid

Purity: 96% CAS No.: 66990-30-5 Molecular Formula: CH3(CH2)9C≡CC≡C(CH2)8CO2H Formula Weight: 346.56 Melting Point: 55-59° Beilstein Reference: 1880414

Formula: C₂₃H₃₈O₂
MW: 346.56 g/mol
Melting Pt: 55…59 °C
Storage Temperature: Ambient
MDL Number: MFCD00041683
CAS Number: 66990-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR