Naringin hydrate 98%
Supplier: Thermo Scientific Chemicals
L10163-09
L10163-18
CAAAAL10163-09EA
37.76
CAD
CAAAAL10163-09
CAAAAL10163-18
Naringin hydrate 98%
Naringin hydrate
A citrus bioflavanoid that inhibits cytochrome P450 monooxygenase activity
Purity: 98%
CAS No.: 10236-47-2
Molecular Formula: C27H32O14.xH2O
Formula Weight: 580.55(anhy)
Melting Point: ca 166° dec.
Beilstein Reference: 102012
Optical Rotation: -90° (c=1 in ethanol)
MDL No.: MFCD00149445
Formula:
C₂₇H₃₄O₁₅ Melting Pt: ca 166 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00149445 CAS Number: 132203-74-8 EINECS: 233-566-4 Merck Index: 14,6424 |
Specification Test Results
Optical rotation | -90° (c=1 in ethanol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...