Benzofuroxan 98%

Supplier: Thermo Scientific Chemicals

L10144-09 L10144-18
CAAAAL10144-09EA 91.8 CAD
CAAAAL10144-09 CAAAAL10144-18
Benzofuroxan 98%
Benzofuroxan

Purity: 98% CAS No.: 480-96-6 Molecular Formula: C6H4N2O2 Formula Weight: 136.11 Melting Point: 67-71° Beilstein Reference: 125112 MDL No.: MFCD00005807

Formula: C₆H₄N₂O₂
Melting Pt: 67…71 °C
Storage Temperature: Ambient
MDL Number: MFCD00005807
CAS Number: 480-96-6
EINECS: 207-559-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR