2-[(2-Methylphenoxy)methyl]oxirane ≥85%, tech.

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-((o-Tolyloxy)methyl)oxirane

L10136-14 L10136-22
CAAAAL10136-14EA 48.51 CAD
CAAAAL10136-14 CAAAAL10136-22
2-[(2-Methylphenoxy)methyl]oxirane ≥85%, tech.
2-[(2-Methylphenoxy)methyl]oxirane

Purity: 85% CAS No.: 2210-79-9 Molecular Formula: C10H12O2 Formula Weight: 164.21 Flash Point: 120°(248°F) Refractive Index: 1.5290 Boiling Point: 119-120°/6mm Beilstein Reference: 4585 UN No.: 3082 MDL No.: MFCD00037847

Formula: C₁₀H₁₂O₂
MW: 164.21 g/mol
Boiling Pt: 119…120 °C/6 mm
Density: 1.079
Flash Pt: 120 °C (248 °F)
Storage Temperature: Ambient
MDL Number: MFCD00037847
CAS Number: 2210-79-9
EINECS: 218-645-3
UN: 3082
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR