3'-Isothiocyanatoacetophenone 97%

Supplier: Thermo Scientific Chemicals
Danger

L10134-03 L10134-06
CAAAAL10134-03EA 42.03 CAD
CAAAAL10134-03 CAAAAL10134-06
3'-Isothiocyanatoacetophenone 97%
3'-Isothiocyanatoacetophenone

Purity: 97% CAS No.: 3125-71-1 Molecular Formula: C9H7NOS Formula Weight: 177.23 Flash Point: 110°(230°F) Refractive Index: 1.6480 Odor: Irritating Melting Point: ca 24° Boiling Point: 135-136°/2mm Beilstein Reference: 2803021 UN No.: 2923 MDL No.: MFCD00041081

Formula: C₉H₇NOS
Boiling Pt: 135…136 °C/2 mm
Melting Pt: ca 24 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041081
CAS Number: 3125-71-1
UN: 2923
ADR: "8",III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR