Methylhydrazine sulfate 98%

Supplier: Thermo Scientific Chemicals
Danger

L10122-06 L10122-14
CAAAAL10122-06EA 189.6 CAD
CAAAAL10122-06 CAAAAL10122-14
Methylhydrazine sulfate 98%
Methylhydrazine sulfate

Purity: 98% CAS No.: 302-15-8 Molecular Formula: CH3NHNH2.H2SO4 Formula Weight: 144.15 Melting Point: 141-143° Beilstein Reference: 3693575 UN No.: 2811 MDL No.: MFCD00035423

Formula: CH₆N₂
Melting Pt: 141…143 °C
Storage Temperature: Ambient
MDL Number: MFCD00035423
CAS Number: 302-15-8
EINECS: 206-115-4
UN: 2811
ADR: "6.1",III
Merck Index: 14,6086

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR