Phenyl trifluoromethyl sulfide 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Phenyl(trifluoromethyl)sulfane, Phenyl(trifluoromethyl)sulphane, alpha,alpha,alpha-Trifluorothioanisole, (Trifluoromethylthio)benzene

L10114-03 L10114-06
CAAAAL10114-03EA 49.93 CAD
CAAAAL10114-03 CAAAAL10114-06
Phenyl trifluoromethyl sulfide 98%
Phenyl trifluoromethyl sulfide

Purity: 98% CAS No.: 456-56-4 Molecular Formula: C7H5F3S Formula Weight: 178.18 Flash Point: 37°(98°F) Refractive Index: 1.4655 Odor: Unpleasant Boiling Point: 140-142° Beilstein Reference: 2043134 UN No.: 1993 MDL No.: MFCD00040839

Formula: C₇H₅F₃S
MW: 178.17 g/mol
Boiling Pt: 140…142 °C
Density: 1.249
Flash Pt: 37 °C (98 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040839
CAS Number: 456-56-4
EINECS: 207-270-0
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR