(S)-2,4-Diaminobutanoic acid dihydrochloride ≥98% may cont. up to ca 10% monohydrochloride
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
L-2,4-Diaminobutanoic acid dihydrochloride, H-Dab-OH 2HCl, L-2,4-Diaminobutyric acid dihydrochloride, (2S)-2,4-Diaminobutanoic acid dihydrochloride, H-Daba-OH·2HCl, (S)-(+)-2,4-Diaminobutyric acid dihydrochloride
L10093-03
L10093-06
CAAAAL10093-03EA
122.61
CAD
CAAAAL10093-03
CAAAAL10093-06
(S)-2,4-Diaminobutanoic acid dihydrochloride ≥98% may cont. up to ca 10% monohydrochloride
(S)-2,4-Diaminobutanoic acid dihydrochloride
Purity: 98+%
CAS No.: 1883-09-6
Molecular Formula: C4H10N2O2.2HCl
Formula Weight: 191.06
Melting Point: ca 200° dec.
Beilstein Reference: 5763078
Optical Rotation: +18° (c=1 in 5N HCl)
MDL No.: MFCD00064561
Formula:
C₄H₁₂Cl₂N₂O₂ MW: 191.06 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00064561 CAS Number: 1883-09-6 EINECS: 217-542-0 |
Specification Test Results
Optical rotation | +18° (c=1 in 5N HCl) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...