3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carboxylic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L10092-09
L10092-18
CAAAAL10092-09EA
118.97
CAD
CAAAAL10092-09
CAAAAL10092-18
3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carboxylic acid 99%
3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carboxylic acid
Purity: 99%
CAS No.: 3919-74-2
Molecular Formula: C11H7ClFNO3
Formula Weight: 255.63
Melting Point: 201-203°
Beilstein Reference: 806523
Formula:
C₁₁H₇ClFNO₃ Melting Pt: 201…203 °C Storage Temperature: Ambient |
MDL Number:
MFCD00055659 CAS Number: 3919-74-2 EINECS: 223-486-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...