Propargyl acetate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L10031-06
L10031-14
CAAAAL10031-06EA
64.31
CAD
CAAAAL10031-06
CAAAAL10031-14
Propargyl acetate 97%
Propargyl acetate
Purity: 97%
CAS No.: 627-09-8
Molecular Formula: C5H6O2
Formula Weight: 98.10
Flash Point: 31°(88°F)
Refractive Index: 1.4185
Odor: Characteristic
Boiling Point: 124°
Beilstein Reference: 1742046
UN No.: 1992
MDL No.: MFCD00041601
Formula:
C₅H₆O₂ Boiling Pt: 124 °C Density: 0.989 Flash Pt: 31 °C(88 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00041601 CAS Number: 627-09-8 UN: 1992 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...