3',5'-Dichloro-2'-hydroxyacetophenone 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L10006-06
L10006-14
CAAAAL10006-06EA
83.9
CAD
CAAAAL10006-06
CAAAAL10006-14
3',5'-Dichloro-2'-hydroxyacetophenone 99%
3',5'-Dichloro-2'-hydroxyacetophenone
Purity: 99%
CAS No.: 3321-92-4
Molecular Formula: C8H6Cl2O2
Formula Weight: 205.04
Melting Point: 95-98°
Boiling Point: 132-134°/18mm
MDL No.: MFCD00016417
Formula:
C₈H₆Cl₂O₂ MW: 205.04 g/mol Boiling Pt: 132…134 °C/18 mm Melting Pt: 95…98 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016417 CAS Number: 3321-92-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...