Methyl-2-naphthoate 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
Methyl 2-naphthoate
L09973-03
L09973-06
CAAAAL09973-03EA
53.25
CAD
CAAAAL09973-03
CAAAAL09973-06
Methyl-2-naphthoate 99%
Methyl 2-naphthoate
Purity: 99%
CAS No.: 2459-25-8
Molecular Formula: C12H10O2
Formula Weight: 186.21
Melting Point: 75-79°
Boiling Point: 290°
MDL No.: MFCD00017345
Formula:
C₁₂H₁₀O₂ MW: 186.21 g/mol Boiling Pt: 290 °C Melting Pt: 75…79 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017345 CAS Number: 2459-25-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...