Diethyl chlorophosphite 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L09919-06
L09919-14
CAAAAL09919-06EA
70.66
CAD
CAAAAL09919-06
CAAAAL09919-14
Diethyl chlorophosphite 97%
Diethyl chlorophosphite
Fieser: 1,248 20,142
Purity: 97%
CAS No.: 589-57-1
Molecular Formula: C4H10ClO2P
Formula Weight: 156.55
Flash Point: 25°(77°F)
Refractive Index: 1.4375
Odor: Acrid
Boiling Point: 153-155°
Beilstein Reference: 1098392
Merck Reference: 14,3841
UN No.: 2920
MDL No.: MFCD00009074
Formula:
C₄H₁₀ClO₂P Boiling Pt: 153…155 °C Density: 1.082 Flash Pt: 25 °C(77 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009074 CAS Number: 589-57-1 EINECS: 209-652-2 UN: 2920 ADR: "8",II Merck Index: 14,3841 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...