N-Boc-L-prolinol 98+%
Supplier: Thermo Scientific Chemicals
Synonyms:
N-(tert-Butoxycarbonyl)-L-prolinol, 1-Boc-2-(S)-pyrrolidinemethanol, (S)-1-(tert-Butoxycarbonyl)-2-pyrrolidinemethanol, (S)-1-Boc-2-pyrrolidinemethanol, N-Boc-L-Prolinol
L09885-03
L09885-06
L09885-14
CAAAAL09885-03EA
77.1
CAD
CAAAAL09885-03
CAAAAL09885-MD
CAAAL09885-06
CAAAL09885-14
N-Boc-L-prolinol 98+%
N-Boc-L-prolinol
Purity: 98+%
CAS No.: 69610-40-8
Molecular Formula: C10H19NO3
Formula Weight: 201.27
Storage Temperature: 2°C to 8°C
Melting Point: 59-62°
Beilstein Reference: 3542667
Optical Rotation: -54° (c=5 in methanol)
MDL No.: MFCD00066232
Formula:
C₁₀H₁₉NO₃ Melting Pt: 59…62 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00066232 CAS Number: 69610-40-8 |
Specification Test Results
Optical rotation | -54° (c=5 in methanol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...