2-Tetrahydrofurfuryl isothiocyanate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-(Isothiocyanatomethyl)tetrahydrofuran
L09878-03
L09878-06
CAAAAL09878-03EA
65.73
CAD
CAAAAL09878-03
CAAAAL09878-06
2-Tetrahydrofurfuryl isothiocyanate 98%
2-Tetrahydrofurfuryl isothiocyanate
Purity: 98%
CAS No.: 36810-87-4
Molecular Formula: C6H9NOS
Formula Weight: 143.21
Storage Temperature: 2°C to 8°C
Refractive Index: 1.5350
Boiling Point: 122-125°/18mm
Beilstein Reference: 111696
UN No.: 2922
MDL No.: MFCD00022476
Formula:
C₆H₉NOS MW: 143.21 g/mol Boiling Pt: 122…125 °C/18 mm Density: 1.120 Storage Temperature: Refrigerator |
MDL Number:
MFCD00022476 CAS Number: 36810-87-4 UN: 2922 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...