5-Cyano-2-methylpyridine 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 6-Methylnicotinonitrile, 5-Cyano-2-picoline

L09875-03 L09875-06
CAAAAL09875-03EA 50.4 CAD
CAAAAL09875-03 CAAAAL09875-06
5-Cyano-2-methylpyridine 99%
5-Cyano-2-methylpyridine

Purity: 99% CAS No.: 3222-48-8 Molecular Formula: C7H6N2 Formula Weight: 118.14 Melting Point: 83-87° Boiling Point: 216-217° MDL No.: MFCD00038042

Formula: C₇H₆N₂
Boiling Pt: 216…217 °C
Melting Pt: 83…87 °C
Storage Temperature: Ambient
MDL Number: MFCD00038042
CAS Number: 3222-48-8
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR