1,1,1-Trifluoro-2,3-epoxypropane 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3,3,3-Trifluoro-1,2-propenoxide
L09869-03
L09869-06
CAAAAL09869-03EA
137.3
CAD
CAAAAL09869-03
CAAAAL09869-06
1,1,1-Trifluoro-2,3-epoxypropane 98%
1,1,1-Trifluoro-2,3-epoxypropane
Purity: 98%
CAS No.: 359-41-1
Molecular Formula: C3H3F3O
Formula Weight: 112.05
Storage Temperature: 2°C to 8°C
Flash Point: -40°(-40°F)
Refractive Index: 1.297
Boiling Point: 38-40°
Beilstein Reference: 103730
UN No.: 1992
MDL No.: MFCD00041506
Formula:
C₃H₃F₃O MW: 112.05 g/mol Boiling Pt: 38…40 °C Density: 1.300 Flash Pt: -40 °C(-40 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00041506 CAS Number: 359-41-1 UN: 1992 ADR: "3",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...