4'-Fluoro-2'-methoxyacetophenone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(4-Fluoro-2-methoxyphenyl)ethanone

L09862-03 L09862-06
CAAAAL09862-03EA 138.25 CAD
CAAAAL09862-03 CAAAAL09862-06
4'-Fluoro-2'-methoxyacetophenone 97%
4'-Fluoro-2'-methoxyacetophenone

Purity: 97% CAS No.: 51788-80-8 Molecular Formula: C9H9FO2 Formula Weight: 168.17 Melting Point: 51-53° Beilstein Reference: 2251986

Formula: C₉H₉FO₂
MW: 168.17 g/mol
Melting Pt: 51…53 °C
Storage Temperature: Ambient
MDL Number: MFCD00061143
CAS Number: 51788-80-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR