(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Naproxen, (S)-2-(6-Methoxynaphthalen-2-yl)propanoic acid, (S)-(+)-6-Methoxy-alpha-methyl-2-naphthaleneacetic Acid

L09855-03 L09855-06 L09855-14
CAAAAL09855-03EA 49.61 CAD
CAAAAL09855-03 CAAAAL09855-06 CAAAL09855-14
(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid 99%
(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid

Purity: 99% CAS No.: 22204-53-1 Molecular Formula: C14H14O3 Formula Weight: 230.26 Odor: Odorless Melting Point: 154-156° Beilstein Reference: 3591067 Optical Rotation: +66° (c=1 in chloroform) UN No.: 2811 MDL No.: MFCD00010500

Formula: C₁₄H₁₄O₃
MW: 230.26 g/mol
Melting Pt: 154…156 °C
Storage Temperature: Ambient
MDL Number: MFCD00010500
CAS Number: 22204-53-1
EINECS: 244-838-7
UN: 2811
ADR: 6.1 III
Merck Index: 14,6417

Order Now

Specification Test Results

Optical rotation +66° (c=1 in chloroform)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR