DL-3-Phenyllactic acid ≥98%
Supplier: Thermo Scientific Chemicals
L09854-03
L09854-06
CAAAAL09854-03EA
55.14
CAD
CAAAAL09854-03
CAAAAL09854-06
DL-3-Phenyllactic acid ≥98%
DL-3-Phenyllactic acid
Purity: 98+%
CAS No.: 828-01-3
Molecular Formula: C9H10O3
Formula Weight: 166.18
Odor: Odorless
Melting Point: 95-98°
Beilstein Reference: 2209791
MDL No.: MFCD00065928
Formula:
C₉H₁₀O₃ Melting Pt: 95…98 °C Storage Temperature: Ambient |
MDL Number:
MFCD00065928 CAS Number: 828-01-3 EINECS: 212-580-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...