Bromomethyl methyl ether tech. 90%, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Warning
|
L09837-06
L09837-14
CAAAAL09837-06EA
66.52
CAD
CAAAAL09837-06
CAAAAL09837-14
Bromomethyl methyl ether tech. 90%, Technical Grade
Bromomethyl methyl ether
Fieser: 10,56
Purity: 90%
CAS No.: 13057-17-5
Molecular Formula: BrCH2OCH3
Formula Weight: 124.97
Storage Temperature: 2°C to 8°C
Flash Point: 26°(78°F)
Refractive Index: 1.4520-1.4600
Odor: Pungent
Boiling Point: 86-87°
Beilstein Reference: 1730853
UN No.: 1992
MDL No.: MFCD00000171
Formula:
C₂H₅BrO Boiling Pt: 86…87 °C Density: 1.531 Flash Pt: 26 °C(78 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000171 CAS Number: 13057-17-5 UN: 1992 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...