Zinc-copper couple (∼1 - 3% Cu)
Supplier: Thermo Scientific Chemicals
|
Warning
|
L09811-14
L09811-22
CAAAAL09811-14EA
149.78
CAD
CAAAAL09811-14
CAAAAL09811-22
Zinc-copper couple (∼1 - 3% Cu)
Zinc-copper couple
Fieser: 1,1292 13,348 15,367 16,387 17,407 18,408 19,408 21,492
CAS No.: 53801-63-1
Molecular Formula: Zn/Cu
Storage Temperature: 2°C to 8°C
Melting Point: 419-420°
UN No.: 3089
MDL No.: MFCD00084851
Formula:
CuZn MW: 128.926 g/mol Melting Pt: 419…420 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00084851 CAS Number: 53801-63-1 EINECS: 684-244-9 UN: 3089 ADR: 4.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...