2-Chloroethyl methacrylate 97% stabilized
Supplier: Thermo Scientific Chemicals
|
Danger
|
Stabilization:
Stabilised with hydroquinone (HQ) 400 - 600 ppm
L09783-09
L09783-18
CAAAAL09783-09EA
172.22
CAD
CAAAAL09783-09
CAAAAL09783-18
2-Chloroethyl methacrylate 97% stabilized
2-Chloroethyl methacrylate
Stab. with 500ppm hydroquinone
Purity: 97%
CAS No.: 1888-94-4
Molecular Formula: C6H9ClO2
Formula Weight: 148.59
Flash Point: 69°(156°F)
Refractive Index: 1.4510
Boiling Point: 170°
Beilstein Reference: 956599
UN No.: 2810
MDL No.: MFCD00045291
Formula:
C₆H₉ClO₂ Boiling Pt: 170 °C Flash Pt: 69 °C(156 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00045291 CAS Number: 1888-94-4 EINECS: 217-566-1 UN: 2810 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...