(S)-Methyl-2-amino-6-(((benzyloxy)carbonyl)amino)hexanoate hydrochloride 95%
Supplier: Thermo Scientific Chemicals
L09744-03
L09744-06
CAAAAL09744-03EA
51.51
CAD
CAAAAL09744-03
CAAAAL09744-06
(S)-Methyl-2-amino-6-(((benzyloxy)carbonyl)amino)hexanoate hydrochloride 95%
(S)-Methyl-2-amino-6-(((benzyloxy)carbonyl)amino)hexanoate hydrochloride
Purity: 95%
CAS No.: 27894-50-4
Molecular Formula: C15H22N2O4.HCl
Formula Weight: 330.81
Storage Temperature: 2°C to 8°C
Melting Point: 115-118°
Beilstein Reference: 3578476
Optical Rotation: +14.5° (c=2 in water)
MDL No.: MFCD00034846
Formula:
C₁₅H₂₃ClN₂O₄ Melting Pt: 115…118 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00034846 CAS Number: 27894-50-4 EINECS: 248-715-9 |
Specification Test Results
Optical rotation | +14.5° (c=2 in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...