trans-2-Butenyl acetate 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L09734-06
L09734-14
CAAAAL09734-06EA
172.22
CAD
CAAAAL09734-06
CAAAAL09734-14
trans-2-Butenyl acetate 95%
trans-2-Butenyl acetate
Purity: 95%
CAS No.: 628-08-0
Molecular Formula: C6H10O2
Formula Weight: 114.15
Flash Point: 31°(87°F)
Refractive Index: 1.4200
Boiling Point: 128-130°
Beilstein Reference: 1720994
UN No.: 3272
MDL No.: MFCD00039912
Formula:
C₆H₁₀O₂ Boiling Pt: 128…130 °C Density: 0.919 Flash Pt: 31 °C (87 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00039912 CAS Number: 628-08-0 UN: 3272 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...