9-Ethynyl-9-fluorenol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L09717-03
L09717-06
CAAAAL09717-03EA
64.62
CAD
CAAAAL09717-03
CAAAAL09717-06
9-Ethynyl-9-fluorenol 98%
9-Ethynyl-9-fluorenol
Purity: 98%
CAS No.: 13461-74-0
Molecular Formula: C15H10O
Formula Weight: 206.25
Melting Point: 109-110°
Beilstein Reference: 2104685
Formula:
C₁₅H₁₀O Melting Pt: 109…110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041555 CAS Number: 13461-74-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...