n-Propyl acrylate 95% stabilised

Supplier: Thermo Scientific Chemicals
Warning


Stabilization: 0.1% Stabilised with 4-methoxyphenol (MEHQ)

L09714-09 L09714-18
CAAAAL09714-09EA 110.44 CAD
CAAAAL09714-09 CAAAAL09714-18
n-Propyl acrylate 95% stabilised
n-Propyl acrylate

Stab. with 0.1% 4-methoxyphenol


Purity: 95% CAS No.: 925-60-0 Molecular Formula: CH2=CHCO2(CH2)2CH3 Formula Weight: 114.15 Flash Point: 24°(75°F) Refractive Index: 1.4123 Boiling Point: 43-44°/40mm Beilstein Reference: 1744954 UN No.: 3272 MDL No.: MFCD00048693

Formula: C₆H₁₀O₂
MW: 114.144 g/mol
Boiling Pt: 43…44 °C (40 mmHg)
Density: 0.920
Flash Pt: 24 °C (75 °F)
Storage Temperature: Ambient
MDL Number: MFCD00048693
CAS Number: 925-60-0
EINECS: 213-120-5
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR