Fast blue BB salt
Supplier: Thermo Scientific Chemicals
|
Warning
|
L09704-03
L09704-06
L09704-14
CAAAAL09704-03EA
47.87
CAD
CAAAAL09704-03
CAAAAL09704-06
CAAAAL09704-14
Fast blue BB salt
Fast blue BB salt
CAS No.: 5486-84-0
Molecular Formula: C34H36Cl4N6O6Zn
Formula Weight: 831.89
Storage Temperature: -30°C to -10°C
Odor: Odorless
Melting Point: ca 157° dec.
Beilstein Reference: 1845288
MDL No.: MFCD00074765
Formula:
C₃₄H₃₆Cl₄N₆O₆Zn Melting Pt: ca 157 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00074765 CAS Number: 5486-84-0 EINECS: 226-817-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...