4-(2-Pyridylazo)resorcinol 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L09667-03
L09667-06
CAAAAL09667-03EA
47.56
CAD
CAAAAL09667-03
CAAAAL09667-06
4-(2-Pyridylazo)resorcinol 99%
4-(2-Pyridylazo)resorcinol
Purity: 99%
CAS No.: 1141-59-9
Molecular Formula: C11H9N3O2
Formula Weight: 215.21
Melting Point: ca 187° dec.
Beilstein Reference: 184665
Formula:
C₁₁H₉N₃O₂ Melting Pt: ca 187 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00044237 CAS Number: 1141-59-9 EINECS: 214-528-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...