2,3,4,5-Tetramethyl-2-cyclopentenone cis- and trans-mixture 95%
Supplier: Thermo Scientific Chemicals
Warning
|
L09617-06
L09617-14
CAAAAL09617-06EA
116.76
CAD
CAAAAL09617-06
CAAAAL09617-14
2,3,4,5-Tetramethyl-2-cyclopentenone cis- and trans-mixture 95%
2,3,4,5-Tetramethyl-2-cyclopentenone
Purity: 95%
CAS No.: 54458-61-6
Molecular Formula: C9H14O
Formula Weight: 138.21
Flash Point: 73°(163°F)
Refractive Index: 1.4760
Boiling Point: 99-100°/30mm
Beilstein Reference: 2324088
MDL No.: MFCD00010248
Formula:
C₉H₁₄O Boiling Pt: 99…100 °C/30 mm Density: 0.917 Flash Pt: 73 °C (163 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00010248 CAS Number: 54458-61-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...