Ethyl-3-isocyanatopropionate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L09616-03
L09616-06
CAAAAL09616-03EA
77.89
CAD
CAAAAL09616-03
CAAAAL09616-06
Ethyl-3-isocyanatopropionate 98%
Ethyl-3-isocyanatopropionate
Purity: 98%
CAS No.: 5100-34-5
Molecular Formula: OCNCH2CH2CO2CH2CH3
Formula Weight: 143.14
Storage Temperature: 2°C to 8°C
Flash Point: 92°(197°F)
Refractive Index: 1.4290
Odor: Irritating
Boiling Point: 74-75°/4mm
Beilstein Reference: 1705618
UN No.: 2206
MDL No.: MFCD00078461
Formula:
C₆H₉NO₃ Boiling Pt: 74…75 °C/4 mm Density: 1.087 Flash Pt: 92 °C(197 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00078461 CAS Number: 5100-34-5 UN: 2206 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...