4,4'-Methylenebis(cyclohexylamine) (mixture of isomers) mixture of stereoisomers ≥98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
4,4'-Diaminodicyclohexylmethane, 4,4-Diaminodicyclohexyl methane, 4,4'-Methylenebis(aminocyclohexane), Bis(4-aminocyclohexyl)methane
L09598-14
L09598-22
CAAAAL09598-14EA
64.62
CAD
CAAAAL09598-14
CAAAAL09598-22
4,4'-Methylenebis(cyclohexylamine) (mixture of isomers) mixture of stereoisomers ≥98%
4,4'-Methylenebis(cyclohexylamine) (mixture of isomers)
Purity: 98+%
CAS No.: 1761-71-3
Molecular Formula: C13H26N2
Formula Weight: 210.37
Flash Point: 159°(318°F)
Odor: Amine-like
pH: 11.5
Melting Point: ca 43°
Boiling Point: 325-327°
Beilstein Reference: 2079179
UN No.: 2928
MDL No.: MFCD00001496
Formula:
C₁₃H₂₆N₂ MW: 210.37 g/mol Boiling Pt: 325…327 °C Melting Pt: ca 43 °C Flash Pt: 159 °C(318 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001496 CAS Number: 1761-71-3 EINECS: 217-168-8 UN: 3259 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...