α,2-Dibromo-5-fluorotoluene 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-Bromo-2-(bromomethyl)-4-fluorobenzene, 2-Bromo-5-fluorobenzyl bromide

L09553-03 L09553-06
CAAAAL09553-03EA 118.34 CAD
CAAAAL09553-03 CAAAAL09553-06
α,2-Dibromo-5-fluorotoluene 97%
α,2-Dibromo-5-fluorotoluene

Purity: 97% CAS No.: 112399-50-5 Molecular Formula: C7H5Br2F Formula Weight: 267.92 Refractive Index: 1.5900 Melting Point: 33-36° Boiling Point: 89-90°/1mm Beilstein Reference: 4177658 UN No.: 3265 MDL No.: MFCD00042187

Formula: C₇H₅Br₂F
MW: 267.92 g/mol
Boiling Pt: 89…90 °C/1 mm
Melting Pt: 33…36 °C
Storage Temperature: Ambient
MDL Number: MFCD00042187
CAS Number: 112399-50-5
UN: 3265
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR