2,2,3,4,4,4-Hexafluorobutyl methacrylate 96% stabilized
Supplier: Thermo Scientific Chemicals
|
Warning
|
L09543-14
L09543-22
CAAAAL09543-06EA
78.7
CAD
CAAAAL09543-06
CAAAAL09543-14
CAAAAL09543-22
2,2,3,4,4,4-Hexafluorobutyl methacrylate 96% stabilized
2,2,3,4,4,4-Hexafluorobutyl methacrylate
Purity: 96%
CAS No.: 36405-47-7
Molecular Formula: C8H8F6O2
Formula Weight: 250.14
Storage Temperature: 2°C to 8°C
Flash Point: 56°(132°F)
Refractive Index: 1.3620
Boiling Point: 158°
Beilstein Reference: 2725177
UN No.: 3272
MDL No.: MFCD00042311
Formula:
C₈H₈F₆O₂ Boiling Pt: 158 °C Density: 1.352 Flash Pt: 56 °C(132 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00042311 CAS Number: 36405-47-7 UN: 3272 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...