5-(4-Nitrophenyl)-1H-tetrazole 97%

Supplier: Thermo Scientific Chemicals
Warning

L09531-03 L09531-06
CAAAAL09531-03EA 175.38 CAD
CAAAAL09531-03 CAAAAL09531-06
5-(4-Nitrophenyl)-1H-tetrazole 97%
5-(4-Nitrophenyl)-1H-tetrazole

Purity: 97% CAS No.: 16687-60-8 Molecular Formula: C7H5N5O2 Formula Weight: 191.15 Melting Point: ca 223° dec. Beilstein Reference: 192840 MDL No.: MFCD00068729

Formula: C₇H₅N₅O₂
Melting Pt: ca 223 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00068729
CAS Number: 16687-60-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR