3,4-(Methylenedioxy)phenylacetonitrile 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L09518-09
L09518-18
CAAAAL09518-09EA
107.6
CAD
CAAAAL09518-09
CAAAAL09518-18
3,4-(Methylenedioxy)phenylacetonitrile 98+%
3,4-(Methylenedioxy)phenylacetonitrile
Purity: 98+%
CAS No.: 4439-02-5
Molecular Formula: C9H7NO2
Formula Weight: 161.16
Flash Point:
Formula:
C₉H₇NO₂ MW: 161.16 g/mol Boiling Pt: 135…140 °C/5 mm Melting Pt: 41…45 °C Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005835 CAS Number: 4439-02-5 EINECS: 224-655-9 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...