Vinyl chloroacetate 99% stabilized

Supplier: Thermo Scientific Chemicals
Danger


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

L09446-09 L09446-18
CAAAAL09446-09EA 79.79 CAD
CAAAAL09446-09 CAAAAL09446-18
Vinyl chloroacetate 99% stabilized
Vinyl chloroacetate

Stab. with 4-methoxyphenol


Purity: 99% CAS No.: 2549-51-1 Molecular Formula: ClCH2CO2CH=CH2 Formula Weight: 120.54 Flash Point: 51°(123°F) Refractive Index: 1.4435 Odor: Pungent Boiling Point: 136° Beilstein Reference: 1741679 UN No.: 2589 MDL No.: MFCD00045275

Formula: C₄H₅ClO₂
Boiling Pt: 136 °C
Density: 1.190
Flash Pt: 51 °C (123 °F)
Storage Temperature: Ambient
MDL Number: MFCD00045275
CAS Number: 2549-51-1
EINECS: 219-834-3
UN: 2589
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR