Cyclopropylacetic acid 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L09416-03
L09416-06
CAAAAL09416-03EA
133.98
CAD
CAAAAL09416-03
CAAAAL09416-06
Cyclopropylacetic acid 97%
Cyclopropylacetic acid
Purity: 97%
CAS No.: 5239-82-7
Molecular Formula: C5H8O2
Formula Weight: 100.12
Flash Point: 93°(199°F)
Refractive Index: 1.4350
Odor: Pungent
Boiling Point: 189-191°
Beilstein Reference: 2323311
UN No.: 3265
MDL No.: MFCD00041544
Formula:
C₅H₈O₂ Boiling Pt: 189…191 °C Density: 1.076 Flash Pt: 93 °C(199 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00041544 CAS Number: 5239-82-7 UN: 3265 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...