cis-4-Cyclohexene-1,2-dicarboxylic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (1R,2S)-rel-Cyclohex-4-ene-1,2-dicarboxylic acid

L09392-14 L09392-22
CAAAAL09392-14EA 89.9 CAD
CAAAAL09392-14 CAAAAL09392-22 CAAAAL09392-36
cis-4-Cyclohexene-1,2-dicarboxylic acid 98%
cis-4-Cyclohexene-1,2-dicarboxylic acid

Purity: 98% CAS No.: 2305-26-2 Molecular Formula: C8H10O4 Formula Weight: 170.17 Melting Point: 165-170° Beilstein Reference: 2050419

Formula: C₈H₁₀O₄
MW: 170.17 g/mol
Melting Pt: 165…170 °C
Storage Temperature: Ambient
MDL Number: MFCD00070482
CAS Number: 2305-26-2
EINECS: 218-974-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR